Cabozantinib ≥99% (durch HPLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme:
N-(4-((6,7-Dimethoxyquinolin-4-yl)oxy)phenyl)-N-(4-fluorophenyl)cyclopropane-1,1-dicarboxamide, XL184
ENZCHM1820010
ENZOENZCHM1820010EA
206
EUR
ENZOENZCHM1820010
Cabozantinib ≥99% (durch HPLC)
Cabozantinib
Potent inhibitor of VEGFR-2, MET, Kit and FLT3-ITD
Cabozantinib is a very potent inhibitor, with an IC50 of less than 10 nM for VEGFR2, MET, Kit and FLT3-ITD.
Formel:
C₂₈H₂₄FN₃O₅ Molecular Weight: 501,5 g/mol |
MDL:
MFCD20926324 CAS-Nummer: 849217-68-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...