Amp'd® ELISA signal amplification kit
Lieferant: ENZO LIFE SCIENCES
Amp'd® ELISA Signal Amplification Kit provides up to 50-fold increase in sensitivity over traditional ELISAs while detecting lower concentrations of target in samples.
For the amplification of ELISA sensitivity designed to replace traditional alkaline phosphatase (AP) substrates.
The Amp'd® ELISA Signal Amplification kit is designed to replace traditional alkaline phosphatase (AP) substrates, such as pNPP (p-Nitrophenyl phosphate), with a combination substrate and amplifier system that results in greater sensitivity when compared to a classic substrate enzyme linked-immunosorbent assay (ELISA).
Bestellinformation: Contains Signal Amplification Substrate, Amplifier, Substrate Diluent, and Amplifier Diluent.