Amp'd® glp-1 elisa kit, grow cells
Lieferant: ENZO LIFE SCIENCES
Ultra-sensitive AMP'D® GLP-1 ELISA kit enabling the ability to use less sample to detect levels of human Glucagon-like peptide 1 (7-36) amide, a potent promoter of insulin secretion, a major incretin hormone, and therapeutic for type 2 diabetes.
- Ultra-sensitive measurement of GLP-1, detecting as little as 5,5 pg/ml
- Negligible reactivity from similar GLP-1 forms
- High throughput format with results in three hours for up to 40 samples in duplicate
- Fully quantitative results that surpass semi-quantitative Western blot analysis
For the quantitative determination of Glucagon-like peptide 1 (GLP-1) (7-36) amide in serum and plasma of human origin.
The AMP'D® GLP-1 ELISA kit is a colorimetric immunometric enzyme immunoassay kit with results in three hours.
Bestellinformation: Contains Microtiter Plate, Assay Buffer 13, Standard, Wash Buffer Concentrate, Antibody, Conjugate, and AMP'D® (Signal Amplification) Substrate, Amplifier, Substrate Diluent, Amplifier Diluent, and Stop Solution.