E. coli host cell protein ELISA kit
Lieferant: ENZO LIFE SCIENCES
Sensitive (30 ng/ml) ELISA for the quantitative determination of host cell protein contamination in bulk products expressed in E. coli expression systems.
For the quantitative determination of contamination with E.coli host cell protein.
The E. coli host cell protein ELISA kit is a complete, colorimetric, immunometric immunoassay kit for the quantitative determination of E. coli host cell protein contamination with results in 3 hours
Bestellinformation: Contains 1 x 96 well plate, protein standards, 5x dilution buffer, 10x PBS-T, reporting antibody, streptavidin-HRP conjugate, TMB substrate, stop solution, plate sealer.