CHO host cell protein ELISA kit
Lieferant: ENZO LIFE SCIENCES
ENZKIT1280001
ENZOENZKIT1280001EA 1100 EUR
ENZOENZKIT1280001
CHO host cell protein ELISA kit
Tests ELISAs
Sensitive (10 ng/ml) ELISA for the quantitative determination of host cell protein contamination in bulk products expressed in Chinese Hamster Ovary (CHO) expression systems.
For the quantitative determination of contamination with CHO host cell protein.
The CHO host cell protein ELISA kit is a complete, colorimetric, immunometric immunoassay kit for the quantitative determination of Chinese Hamster Ovary (CHO) host cell protein contamination with results in 3 hours
Bestellinformation: Contains 1×96 well plate, protein standards, 5x dilution buffer, 10x PBS-T, reporting antibody, streptavidin-HRP conjugate, TMB substrate, stop solution, plate sealer.