TGF-α ELISA kit, Enzo Life Sciences
Lieferant: ENZO LIFE SCIENCES
Highly sensitive ELISA kit enabling detection of TGF-α in 3,5 hours.
- Sensitive measurement of TGF-α, detecting as little as 9,375 pg/ml
- Negligible reactivity from similar growth factors
- High throughput format with results in 3,5 hours
- Fully quantitative results that surpass semi-quantitative Western blot analysis
For the quantitative determination of TGF-α in serum, plasma, cell culture supernatant, tissue homogenates in human samples.
The TGF-α ELISA kit is a colorimetric immunometric immunoassay kit with results in 3,5 hours.
Bestellinformation: Contains Microtiter Plate, Standard, Detection Antibody, Detection Antibody Diluent, Sample Diluent, Wash Buffer Concentrate, Conjugate, Conjugate Diluent, Substrate, and Stop Solution.