Peptide YY ELISA kit
Lieferant: ENZO LIFE SCIENCES
Highly sensitive Peptide YY ELISA kit enabling detection of PYY in serum in 3,5 hrs.
- Sensitive measurement of PYY, detecting as little as 18,75 pg/ml
- Negligible reactivity from similar peptides
- High throughput format with results in 3,5 hrs
- Fully quantitative results that surpass semi-quantitative Western blot analysis
For the quantitative determination of Peptide YY in serum, plasma, cell culture supernatant and tissue homogenate in human samples.
The Peptide YY ELISA kit is a colorimetric immunometric immunoassay kit with results in 3,5 hours.
Bestellinformation: Contains Microtiter Plate, Standard, Sample Diluent, Detection Antibody, Wash Buffer Concentrate, Conjugate, Conjugate Diluent, Substrate, and Stop Solution.