Peptide YY ELISA kit
Lieferant: ENZO LIFE SCIENCES
ENZKIT1330001
ENZOENZKIT1330001EA 901 EUR
ENZOENZKIT1330001
Peptide YY ELISA kit
Tests ELISAs
Highly sensitive Peptide YY ELISA kit enabling detection of PYY in serum in 3,5 hrs.
  • Sensitive measurement of PYY, detecting as little as 18,75 pg/ml
  • Negligible reactivity from similar peptides
  • High throughput format with results in 3,5 hrs
  • Fully quantitative results that surpass semi-quantitative Western blot analysis
For the quantitative determination of Peptide YY in serum, plasma, cell culture supernatant and tissue homogenate in human samples.
The Peptide YY ELISA kit is a colorimetric immunometric immunoassay kit with results in 3,5 hours.
Bestellinformation: Contains Microtiter Plate, Standard, Sample Diluent, Detection Antibody, Wash Buffer Concentrate, Conjugate, Conjugate Diluent, Substrate, and Stop Solution.