IL-5 ELISA kit, grow cells
Lieferant: ENZO LIFE SCIENCES
ENZKIT1390001
ENZOENZKIT1390001EA 943 EUR
ENZOENZKIT1390001
IL-5 ELISA kit, grow cells
Tests ELISAs
Highly sensitive IL-5 ELISA kit enabling detection of IL-5 in serum, plasma, urine and cell culture supernates in just 3 hours.
  • Sensitive measurement of IL-5, detecting as little as 2 pg/ml.
  • Negligible reactivity to related proteins.
  • High throughput with results in three hours.
  • Fully quantitative results that pass semi-quantitative Western blot analysis.
For the quantitative determination of IL-5 in cell culture supernates, serum, plasma (Heparin & EDTA), urine, in human samples.
The IL-5 ELISA kit is a colorimetric sandwich immunoassay kit with results in three hours.
Bestellinformation: Contains Microtiter Plate, Standard, Sample Buffer, Wash Buffer Concentrate, Antibody, Antibody Buffer, Conjugate, Substrate, and Stop Solution.