Methotrexate ELISA kit
Lieferant: ENZO LIFE SCIENCES
ENZKIT1420001
ENZOENZKIT1420001EA
735
EUR
ENZOENZKIT1420001
Methotrexate ELISA kit
Tests
ELISAs
Ultra sensitive Methotrexate ELISA kit enabling detection of Methotrexate in serum, plasma, and urine in just 1,5 hours.
- Sensitive measurement of methotrexate, detecting as little as 0,087 ng/ml
- Broad dynamic range suitable for a large variety of samples
- High throughput format with results in 1,5 hours for up to 38 samples in duplicate
- Fully quantitative results that surpass semi-quantitative Western blot analysis
For the quantitative determination of methotrexate in serum, plasma and urine in human, mouse, and rat samples. The Methotrexate ELISA kit is a colorimetric competitive immunoassay kit with results in 1,5 hours.
Bestellinformation: Contains Microtiter Plate, Assay Buffer 13, Standard, Wash Buffer Concentrate, Antibody, Antibody Diluent, Conjugate, Substrate, and Stop Solution.