Total PSA (human) ELISA kit
Lieferant: ENZO LIFE SCIENCES
ENZKIT1460001
ENZOENZKIT1460001EA 605 EUR
ENZOENZKIT1460001
Total PSA (human) ELISA kit
Tests ELISAs
Highly sensitive Total PSA ELISA kit enabling detection of Total PSA in human serum, plasma, urine, and tissue culture media samples in just 2,5 hours.
  • Highly-sensitive measurement of Total PSA, detecting as little as 0,287pM (or 0,024ng/ml) with negligible cross reactivity with similar proteins (KLK2, ACT)
  • Compatible with various sample types (plasma, serum, urine, tissue culture media)
  • High throughput format with results in 2,5 hours for up to 40 samples in duplicate
  • Fully quantitative results that surpass semi-quantitative Western blot analysis
For the quantitative determination of Total PSA in human serum, plasma, urine and tissue culture media. The Total PSA (human) ELISA kit is a colorimetric immunometric immunoassay kit with results in 2,5 hours.
Bestellinformation: Contains Microtiter plate, Assay Buffer 13, Standard, Wash Buffer Concentrate, Antibody, Antibody Diluent, Conjugate, Substrate, and Stop Solution.