TIMP-1 (human) ELISA kit
Lieferant: ENZO LIFE SCIENCES
ENZKIT1470001
ENZOENZKIT1470001EA
414
EUR
ENZOENZKIT1470001
TIMP-1 (human) ELISA kit
Tests
ELISAs
Highly sensitive TIMP-1 ELISA kit enabling detection of TIMP-1 in serum, plasma, saliva, urine and tissue culture media in just 2 hours.
- Highly sensitive measurement of TIMP-1, detecting as little as 30 pg/ml
- Negligible cross-reactivity with similar proteins (TIMP-2, TIMP-3, TIMP-4, MMP-1, MMP-2, MMP-3, MMP-7, MMP-8, MMP-9, MMP-13)
- High throughput format with results in two hours for up to 39 samples in duplicate
- Fully quantitative results that surpass semi-quantitative Western blot analysis
For the quantitative determination of TIMP-1 in human serum, plasma, saliva, urine and tissue culture media. The TIMP-1 ELISA kit is a colorimetric immunometric immunoassay kit with results in two hours.
Bestellinformation: Contains Microtiter plate, Assay Buffer 13, Standard, Wash Buffer Concentrate, Antibody, Antibody Diluent, Conjugate, Substrate, and Stop Solution.