Melatonin ELISA kit
Lieferant: ENZO LIFE SCIENCES
ENZKIT1500001
ENZOENZKIT1500001EA 759 EUR
ENZOENZKIT1500001
Melatonin ELISA kit
Tests ELISAs
Ultra-sensitive Melatonin ELISA kit enabling detection of Melatonin in saliva, serum, plasma, and fruit homogenates in just 2 hours.
  • Sensitive measurement of Melatonin, detecting as little as 0,162 ng/ml
  • Broad dynamic range suitable for a large variety of samples
  • High throughput format with results in 2 hours for up to 40 samples in duplicate
  • Validated for a variety of sample matrices including fruit homogenates
For the quantitative determination of melatonin in any species. Validated specifically for saliva, serum, plasma, and fruit samples.
The Melatonin ELISA kit is a colorimetric competitive immunoassay kit with results in 2 hours.
Bestellinformation: Contains Goat anti-mouse IgG Microtiter Plate, Assay Buffer 10, Standard, Wash Buffer Concentrate, Antibody, Tracer, Conjugate, TMB Substrate, and Stop Solution.