VEGF (Human), ELISA kit
Lieferant: ENZO LIFE SCIENCES
Ultra-sensitive ELISA kit for the quantitative detection of human VEGF with 100% cross reactivity to VEGF 165 and negligible cross-reactivity to VEGF 121.
- Sensitive Assay: Detect VEGF as low as 4 pg /ml
- Specific: 100% cross-reactivity to VEGF 165 with negligible cross-reactivity to related molecules including VEGF 121
- Flexible: Wide dynamic range ideal for various matrices
- Higher throughput: Assay up to 38 samples in duplicate with each kit with an assay time of just 2.5 hours
- Efficiency: Color coded reagents help to reduce errors and increase efficiency
For the quantitative determination of human VEGF in serum, plasma, and culture supernatants. The VEGF (human), EIA kit is a sensitive colorimetric immunometric enzyme immunoassay kit with results in just 2,5 hours. Absorbance is read at 450 nm. The wide dynamic range is ideal for various matrices. Assay up to 39 samples in duplicate with each kit.
Bestellinformation: Contains Microtiter Plate, Conjugate, Detector Antibody, Assay buffer, Antibody Diluent, Wash Buffer Concentrate, Standard, TMB Substrate, Stop Solution.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...