AMPIGENE® dNTP Mix
Lieferant: ENZO LIFE SCIENCES
ENZNUC1020500
ENZOENZNUC1020500EA
64.9
EUR
ENZOENZNUC1020500
AMPIGENE® dNTP Mix
Nukleinsäure-Reagenzien
Nukleotide
Ultra-pure, stable, and suitable for a variety of PCR applications.
AMPIGENE® dNTP Mix is an ultrapure mix of deoxyribonucleotide triphosphates to use as building blocks for a wide variety of PCR applications. It can withstand multiple freeze-thaw cycles and shown to retain 95% dNTPs in triphosphate form after 5 weeks at room temperature. AMPIGENE® dNTP Mix has been tested on many molecular biology applications including standard PCR, fast PCR, 1-step PCR, high-fidelity PCR, real-time PCR, lon-range PCR, MDA, RDA, LAMP, and DNA sequencing.