AMPIGENE® Taq DNA polymerase
Lieferant: ENZO LIFE SCIENCES
ENZPRT1000500
ENZOENZPRT1000500EA 165 EUR
ENZOENZPRT1000500
AMPIGENE® Taq DNA polymerase
Nukleinsäure-Reagenzien Enzyme und Kits für Endpunkt-PCR
Optimised standard PCR for enhanced speed, yield, and specificity.
AMPIGENE® Taq DNA Polymerase uses the latest developments in polymerase technology and buffer chemistry to optimize PCR. The AMPIGENE® Taq DNA Polymerase and 5x AMPIGENE® reaction buffer are separate for flexibility and end-user customization. It can be used for routine PCR applications including genotyping, screening, and library construction.
Bestellinformation: Contains AMPIGENE® Taq DNA polymerase (5u/µl)and 5x AMPIGENE® reaction buffer (15mM MgCl2, 5mM dNTPs, enhancers and stabilizers).