Streptavidin (AP conjugate)
Lieferant: ENZO LIFE SCIENCES
ENZPRT1191000
ENZOENZPRT1191000EA 95.7 EUR
ENZOENZPRT1191000
Streptavidin (AP conjugate)
Proteine und Peptide
Native. Purified from Streptomyces avidinii (SA) and calf intestine (AP).
As part of the need to increase the sensitivity of your sandwich ELISA, this streptavidin AP conjugate can be used with your sandwich ELISA that utilizes a biotinylated detector antibody and the AMP'D® ELISA Signal Amplification kit (Prod. no. ENZ-KIT-100) which requires the presence of the AP as the conjugate.