Shrimp recombinant alkaline phosphatase
Lieferant: ENZO LIFE SCIENCES
ENZPRT1611000
ENZOENZPRT1611000EA
186
EUR
ENZOENZPRT1611000
Shrimp recombinant alkaline phosphatase
Proteine und Peptide
Heat-labile alkaline phosphatase for dephosphorylation of DNA, RNA and nucleotides
- Very high specific activity
- Completely heat-inactivated at 65°C
- Removes 5’-phosphates from DNA, RNA, dNTPs and proteins
- Works in restriction enzyme buffers
Pandalus borealis (Northern red shrimp) origin, recombinantly produced in Pichia pastoris.
Shrimp alkaline phosphatase (recombinant), (rSAP) is a multipurpose alkaline phosphatase that is fully inactivated by a short heat treatment. rSAP has all the properties of the native enzyme, but with the additional benefits of greater stability, consistency, and purity.rSAP simplifies cloning and sequencing workflows due to a complete heat inactivation.