PCR workstation, CaptairBio
Lieferant: ERLAB CAPTAIRE
AE206Z00011C21
ERLAAE206Z00011C21EA 3779 EUR
ERLAAE206Z00011C21
PCR workstation, CaptairBio
Dunstabzugshauben und Verkleidungen PCR-Arbeitsstationen und -Abzüge
Designed to perform non-pathogenic handlings, these enclosures has been created for handlings involving a high sample cross contamination risk such as sample preparations before thermocycling, Post PCR DNA sequencing revelation / separation, in vitro fertilization, cellular cultures, vegetal biology, preparation of sterile solutions.
Bitte füllen Sie die Angebotsanfrage aus, damit wir Sie bei der Suche nach dem richtigen Gerät für Ihre Anwendung unterstützen können.
https://de.cmd2.vwr.com/bin/public/fc/main?f=Y5&pimid=26080247
  • The worksurface made of stainless steel makes the cleaning easy
  • Total openings for an easy access into the enclosure
  • Energy efficient
  • Automatic UV cut-off in case of front door opening
  • Internal reflectors: no shadow area
  • Captair® Bio 320 is a non ventilated model
Enclosure design from 800 mm to 1,8 m - equipped with a powerful UV decontamination preventing any handling from cross contamination risks.
Optional work surface in stainless steel 304 l with built-in retention tray or tempered glass work surface with sealed frame and rolling or fixed benches used (these must be ordered separately).
Bitte beachten There is a very wide range of Erlab Fume Hoods and Enclosures. To ensure you receive the correct product for your specific application and setting please complete the contact form as per the url above. Your local Avantor sales representative will then send you an eValiQuest form to get details of your application and requirements. Once we have received this back from you, we will send you a quotation based on your precise requirements ensuring all handling and safety requirements are met. 