Anti-MTR Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB05055
EVBIEB05055EA 339 EUR
EVBIEB05055
Anti-MTR Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MTR (5-methyltetrahydrofolate-homocysteine methyltransferase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Dog, Human, Mouse, Pig, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR