Anti-FYN Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB05153
EVBIEB05153EA 339 EUR
EVBIEB05153
Anti-FYN Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FYN (FYN oncogene related to SRC, FGR, YES)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR