Anti-BAIAP2 (isoform 3) Goat polyclonal antibody unconjugated

Lieferant: Everest Biotech
EB05175
EVBIEB05175EA 339 EUR
EVBIEB05175
Anti-BAIAP2 (isoform 3) Goat polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: BAIAP2 (isoform 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR