Anti-TIRAP Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB05187
EVBIEB05187EA 339 EUR
EVBIEB05187
Anti-TIRAP Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TIRAP (toll-interleukin 1 receptor (TIR) domain containing adaptor protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR