Anti-CHN2 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB05207
EVBIEB05207EA 339 EUR
EVBIEB05207
Anti-CHN2 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CHN2 (chimerin 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR