Anti-B56 beta isoform Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB05241
EVBIEB05241EA 335 EUR
EVBIEB05241
Anti-B56 beta isoform Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: B56 beta isoform
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Dog, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR