Anti-ELMO1 Goat polyclonal antibody unconjugated

Lieferant: Everest Biotech
EB05297
EVBIEB05297EA 339 EUR
EVBIEB05297
Anti-ELMO1 Goat polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: ELMO1 (engulfment and cell motility 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR