Anti-ANXA11 Goat Polyclonal Antibody (Biotin)
Lieferant: Everest Biotech
EB05307-B
EVBIEB05307-BEA 335 EUR
EVBIEB05307-B
Anti-ANXA11 Goat Polyclonal Antibody (Biotin)
Antikörper
Western Blot: Approx 55kDa band observed in Jurkat lysates (calculated MW of 54.4kDa according to NP_001148.1). See non-biotinylated parental product's datasheet for further QC data. Recommended concentration: 1-3µg/ml.
Type: Primary
Antigen: ANXA11
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human