Anti-STUB1 / CHIP Goat Polyclonal Antibody
Lieferant: Everest Biotech
EB05327
EVBIEB05327EA 339 EUR
EVBIEB05327
Anti-STUB1 / CHIP Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: STUB1 / CHIP
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity:
Bestellen