Anti-Peg12 / Frat3 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB05376
EVBIEB05376EA 194 EUR
EVBIEB05376
Anti-Peg12 / Frat3 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Peg12 / Frat3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR