Anti-NOD1 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB05389
EVBIEB05389EA 335 EUR
EVBIEB05389
Anti-NOD1 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NOD1 (nucleotide-binding oligomerization domain containing 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR