Anti-Dicarbonyl Reductase Goat polyclonal antibody unconjugated

Lieferant: Everest Biotech
EB05518
EVBIEB05518EA 339 EUR
EVBIEB05518
Anti-Dicarbonyl Reductase Goat polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: Dicarbonyl Reductase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR