Anti-Dynactin Goat Polyclonal Antibody
Lieferant: Everest Biotech
EB05639
EVBIEB05639EA 335 EUR
EVBIEB05639
Anti-Dynactin Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Dynactin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat