Anti-ANP32B Goat Polyclonal Antibody (Biotinylated)
Lieferant: Everest Biotech
EB05885-B
EVBIEB05885-BEA 335 EUR
EVBIEB05885-B
Anti-ANP32B Goat Polyclonal Antibody (Biotinylated)
Antikörper
Western Blot: Approx 30kDa band observed in Human Peripheral Blood Mononucleocytes lysates (calculated MW of 30.7kDa according to NP_000567.1). Recommended concentration: 1-3µg/ml.
Type: Primary
Antigen: ANP32B
Clonality: Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat, Dog, Pig, Cow