Anti-Pallidin Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB05946
EVBIEB05946EA 339 EUR
EVBIEB05946
Anti-Pallidin Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Pallidin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR