Anti-USP6 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB05974
EVBIEB05974EA 339 EUR
EVBIEB05974
Anti-USP6 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: USP6 (ubiquitin specific peptidase 6 (Tre-2 oncogene))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR