Anti-SMARCE1 Goat Polyclonal Antibody
Lieferant: Everest Biotech
EB06054
EVBIEB06054EA 339 EUR
EVBIEB06054
Anti-SMARCE1 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SMARCE1 (SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily e, member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse