Anti-BOULE / BOLL Goat polyclonal antibody unconjugated

Lieferant: Everest Biotech
EB06074
EVBIEB06074EA 339 EUR
EVBIEB06074
Anti-BOULE / BOLL Goat polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: BOULE / BOLL
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR