Anti-UBA2 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB06098
EVBIEB06098EA 339 EUR
EVBIEB06098
Anti-UBA2 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: UBA2 (ubiquitin-like modifier activating enzyme 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR