Anti-ARAP1 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB06126
EVBIEB06126EA 335 EUR
EVBIEB06126
Anti-ARAP1 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ARAP1 (ArfGAP with RhoGAP domain, ankyrin repeat and PH domain 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR