Anti-CDEP / FARP1 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB06135
EVBIEB06135EA 335 EUR
EVBIEB06135
Anti-CDEP / FARP1 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CDEP / FARP1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR