Anti-PHLDA2 / IPL Goat Polyclonal Antibody
Lieferant: Everest Biotech
EB06184
EVBIEB06184EA 335 EUR
EVBIEB06184
Anti-PHLDA2 / IPL Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PHLDA2 / IPL
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Human