Anti-ACVRL1 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB06267
EVBIEB06267EA 339 EUR
EVBIEB06267
Anti-ACVRL1 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ACVRL1 (activin A receptor type II-like 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR