Anti-USH1C Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB06287
EVBIEB06287EA 339 EUR
EVBIEB06287
Anti-USH1C Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: USH1C (Usher syndrome 1C (autosomal recessive, severe))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR