Anti-PIK3C2A Goat Polyclonal Antibody
Lieferant: Everest Biotech
EB06291
EVBIEB06291EA 339 EUR
EVBIEB06291
Anti-PIK3C2A Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PIK3C2A (phosphatidylinositol-4-phosphate 3-kinase, catalytic subunit type 2 alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Dog, Human, Mouse