Anti-IFT74 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB06339
EVBIEB06339EA 335 EUR
EVBIEB06339
Anti-IFT74 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: IFT74 (intraflagellar transport 74 homolog (Chlamydomonas))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR