Anti-SAMSN1 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB06459
EVBIEB06459EA 335 EUR
EVBIEB06459
Anti-SAMSN1 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SAMSN1 (SAM domain, SH3 domain and nuclear localization signals 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR