Anti-RXR alpha Goat Polyclonal Antibody
Lieferant: Everest Biotech
EB06523
EVBIEB06523EA 335 EUR
EVBIEB06523
Anti-RXR alpha Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RXR alpha
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat