Anti-Swiprosin 1 / EFHD2 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB06552
EVBIEB06552EA 335 EUR
EVBIEB06552
Anti-Swiprosin 1 / EFHD2 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Swiprosin 1 / EFHD2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR