Anti-pan ADH Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB06599
EVBIEB06599EA 335 EUR
EVBIEB06599
Anti-pan ADH Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: pan ADH
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR