Anti-ZDHHC17 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB06609
EVBIEB06609EA 335 EUR
EVBIEB06609
Anti-ZDHHC17 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ZDHHC17 (zinc finger, DHHC-type containing 17)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR