Anti-AKR1C3 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB06637
EVBIEB06637EA 335 EUR
EVBIEB06637
Anti-AKR1C3 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: AKR1C3 (aldo-keto reductase family 1, member C3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR